Learn More
REVMAB BIOSCIENCES USA INC Acetyl-Histone H3 (Lys18) [RM166], Rabbit Monoclonal Antibody
Supplier:  REVMAB BIOSCIENCES USA INC 31105500
Encompass_Preferred
Anti-Acetyl-Histone H3 (Lys18) rabbit monoclonal antibody, Clone RM166 (100UG); WB, ELISA, Multiplex, ChIP, ICC. IHC validated; Product info: www.revmab.com
Catalog No. NC1729092