Learn More
Cell Signaling Technology Acetyl-Histone H3 (Lys9) Matched Antibody Pair, 1 Kit
SDP
Catalog No. 503131891
Encompass
The Acetyl-Histone H3 (Lys9) Matched Antibody Pair is designed for high-throughput immunoassay platforms, including ELISA, MSD, Quanterix Simoa, and Alpha Technology. This pair is optimized for applications requiring customized labeling, such as with biotin, fluorophores, or beads. It contains capture and detection antibodies that recognize non-overlapping epitopes on the H3 (Lys9) acetylate target, providing a versatile solution for studying histone acetylation and chromatin dynamics.
  • Supplied as a 1 kit format containing 100 μg each of Histone H3 (D1H2) and Acetyl-Histone H3 (Lys9) (C5B11) rabbit monoclonal antibodies
  • Formulated in 1X PBS, BSA and azide free
  • Validated for human, mouse, rat, and monkey reactivity
  • Supports high-throughput platforms including ELISA, HTRF, and Luminex
  • Optimal for applications requiring custom antibody conjugation
Catalog No. 50-313-1891 Supplier Cell Signaling Technology Supplier No. 31712S
The Acetyl-Histone H3 (Lys9) Matched Antibody Pair is designed for high-throughput immunoassay platforms, including ELISA, MSD, Quanterix Simoa, and Alpha Technology. This pair is optimized for applications requiring customized labeling, such as with biotin, fluorophores, or beads. It contains capture and detection antibodies that recognize non-overlapping epitopes on the H3 (Lys9) acetylate target, providing a versatile solution for studying histone acetylation and chromatin dynamics.
  • Supplied as a 1 kit format containing 100 μg each of Histone H3 (D1H2) and Acetyl-Histone H3 (Lys9) (C5B11) rabbit monoclonal antibodies
  • Formulated in 1X PBS, BSA and azide free
  • Validated for human, mouse, rat, and monkey reactivity
  • Supports high-throughput platforms including ELISA, HTRF, and Luminex
  • Optimal for applications requiring custom antibody conjugation