Learn More
ABclonal Technology Acetyl-Histone H4-K12
SDP
Catalog No. 5000006052
Encompass_Preferred
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes Two molecules of each of the four core histones (H2A H2B H3 and H4) form an octamer around which approximately 146 bp of DNA is wrapped in repeating units called nucleosomes The linker histone H1 interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures This gene is intronless and encodes a replication-dependent histone that is a member of the histone H4 family Transcripts from this gene lack polyA tails but instead contain a palindromic termination element This gene is found in the large histone gene cluster on chromosome 6
Catalog No. 50-000-06052 Supplier ABclonal Technology Supplier No. A2368320UL
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes Two molecules of each of the four core histones (H2A H2B H3 and H4) form an octamer around which approximately 146 bp of DNA is wrapped in repeating units called nucleosomes The linker histone H1 interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures This gene is intronless and encodes a replication-dependent histone that is a member of the histone H4 family Transcripts from this gene lack polyA tails but instead contain a palindromic termination element This gene is found in the large histone gene cluster on chromosome 6