Learn More
Cell Signaling Technology Acetyl-Histone H4 Matched Antibody Pair, 1 Kit
SDP
Catalog No. 503134398
Encompass
The Acetyl-Histone H4 Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms requiring antibody pairs with custom labeling capabilities. This kit contains capture and detection antibodies that bind to non-overlapping epitopes, making it suitable for applications that require conjugated antibody pairs, such as MSD, Quanterix Simoa, Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF), and Luminex.
  • Contains 100 μg each of Histone H4 mouse monoclonal antibody and Acetylated-Lysine rabbit monoclonal antibody.
  • Supplied in BSA and azide-free formulation for specialized labeling, including fluorophores, lanthanides, biotin, and beads.
  • Validated for reactivity with human, mouse, and monkey samples.
  • Requires storage at -20°C.
Catalog No. 50-313-4398 Supplier Cell Signaling Technology Supplier No. 44076S
The Acetyl-Histone H4 Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms requiring antibody pairs with custom labeling capabilities. This kit contains capture and detection antibodies that bind to non-overlapping epitopes, making it suitable for applications that require conjugated antibody pairs, such as MSD, Quanterix Simoa, Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF), and Luminex.
  • Contains 100 μg each of Histone H4 mouse monoclonal antibody and Acetylated-Lysine rabbit monoclonal antibody.
  • Supplied in BSA and azide-free formulation for specialized labeling, including fluorophores, lanthanides, biotin, and beads.
  • Validated for reactivity with human, mouse, and monkey samples.
  • Requires storage at -20°C.