Learn More
Selleck Chemical LLC Acetyl-L-carnitine
SDP
Catalog No. 5000003048
Encompass_Preferred
O-Acetyl-L-carnitine (Acetyl-L-carnitine O-acetyl-L-carnitine O-Acetylcarnitine) can be synthesis or is naturally found in healthy humans It could help transport fatty acids into the mitochondrial matrix where fatty acid metabolism occurs
Catalog No. 50-000-03048 Supplier Selleck Chemical LLC Supplier No. S550325MG
O-Acetyl-L-carnitine (Acetyl-L-carnitine O-acetyl-L-carnitine O-Acetylcarnitine) can be synthesis or is naturally found in healthy humans It could help transport fatty acids into the mitochondrial matrix where fatty acid metabolism occurs