Learn More
TARGETMOL CHEMICALS INC Acetyl-L-carnitine 1MG
SDP
Catalog No. 502879764
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Acetyl-L-carnitine(RO 447190) is an amino acid that crosses the blood-brain barrier and is often used as a dietary supplement for neuroinflammation.Acetyl-L-carnitine is neuroprotective and can be used in the study of neurological disorders like Alzheimer's disease. Purity 100%
Catalog No. 50-287-9764 Supplier TARGETMOL CHEMICALS INC Supplier No. T265511MG
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Acetyl-L-carnitine(RO 447190) is an amino acid that crosses the blood-brain barrier and is often used as a dietary supplement for neuroinflammation.Acetyl-L-carnitine is neuroprotective and can be used in the study of neurological disorders like Alzheimer's disease. Purity 100%