Learn More
Medchemexpress LLC Acetyl-L-carnitine-d3 hydrochloride | 1334532-17-0 | 99.8% | 25 MG
SDP
Catalog No. 5000450827
Encompass_Preferred
Acetyl-L-carnitine-d3 (O-Acetyl-L-carnitine-d3) hydrochloride is a deuterium-labeled form of Acetyl-L-carnitine hydrochloride. This orally active neuroprotectant and mitochondrial energy metabolism regulator can penetrate the blood-brain barrier. It participates in fatty acid β-oxidation, promotes acetylcholine synthesis, regulates mitochondrial function, and inhibits oxidative stress, enhancing energy metabolism and protecting neurons. It is used in neurodegenerative diseases and metabolic disorder-related diseases.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Enables quantitation during drug development processes.
  • Potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-50827 Supplier Medchemexpress LLC Supplier No. 25MGHYB0762S1
Acetyl-L-carnitine-d3 (O-Acetyl-L-carnitine-d3) hydrochloride is a deuterium-labeled form of Acetyl-L-carnitine hydrochloride. This orally active neuroprotectant and mitochondrial energy metabolism regulator can penetrate the blood-brain barrier. It participates in fatty acid β-oxidation, promotes acetylcholine synthesis, regulates mitochondrial function, and inhibits oxidative stress, enhancing energy metabolism and protecting neurons. It is used in neurodegenerative diseases and metabolic disorder-related diseases.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Enables quantitation during drug development processes.
  • Potential to affect pharmacokinetic and metabolic profiles of drugs.