Learn More
Medchemexpress LLC Acetyl-L-carnitine hydrochloride | 5080-50-2 | MFCD00082230 | >98.0% | 239.70 g/mol | C9H17NO4 · HCl | 10 MG
SDP
Catalog No. 5000296738
Encompass_Preferred
Acetyl-L-carnitine hydrochloride is the hydrochloride salt of O-acetyl-L-carnitine, a mitochondrial energy metabolism regulator and neuroprotective compound used as an analytical standard in biochemical and pharmacological research, particularly for studies of neurodegenerative and metabolic disorders.
  • Hydrochloride salt form that improves stability and solubility.
  • High purity suitable for analytical applications (>98.0%).
  • Molecular weight 239.70 g/mol.
  • Suitable for use as an analytical standard in assays and method validation.
  • Provided in a small package for reference and calibration work.
Catalog No. 50-002-96738 Supplier Medchemexpress LLC Supplier No. HYB0762R10MG
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Acetyl-L-carnitine hydrochloride is the hydrochloride salt of O-acetyl-L-carnitine, a mitochondrial energy metabolism regulator and neuroprotective compound used as an analytical standard in biochemical and pharmacological research, particularly for studies of neurodegenerative and metabolic disorders.
  • Hydrochloride salt form that improves stability and solubility.
  • High purity suitable for analytical applications (>98.0%).
  • Molecular weight 239.70 g/mol.
  • Suitable for use as an analytical standard in assays and method validation.
  • Provided in a small package for reference and calibration work.