Learn More
Cayman Chemical Acetyl-L-Homoserine lactone, Purity: ≥98%, CAS: 51524-71-1, 10 mg
SDP
Catalog No. 502964019
Encompass_Preferred
A C2 N-acyl homoserine lactone; does not induce carbapenem antibiotic biosynthesis in E. carotovora; does not induce fluorescence in E. coli containing a reporter plasmid of GFP in response to exogenous AHLs
Catalog No. 50-296-4019 Supplier Cayman Chemical Supplier No. 2324110 MG
A C2 N-acyl homoserine lactone; does not induce carbapenem antibiotic biosynthesis in E. carotovora; does not induce fluorescence in E. coli containing a reporter plasmid of GFP in response to exogenous AHLs