Learn More
CPC Scientific Ac-His-Ser-Asp-Gly-Ile-Phe-Thr-Asp-Ser-Tyr-Ser-Arg-Tyr-Arg-Lys-Gln-Met-Ala-Val-Lys-Lys-Tyr-Leu-Ala-Ala-Val-Leu-Gly-Lys-Arg-Tyr-Lys-Gln-Arg-Val-Lys-Asn-Lys-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PACA007B
Encompass
ONE-LETTER SEQUENCE: Ac-HSDGIFTDSYSRYRKQMAVKKYLAAVLGKRYKQRVKNK-NH2MOLECULAR FORMULA: C205H333N63O54S1MOLECULAR WEIGHT:4576.36STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [156106-32-0], RESEARCH AREA: HormonalREFERENCES: A.Ravni et al., J. Neurochem., 98, 321 (2006); S.Bourgault et al., Peptides 2006, Proceedings of the 29th European Peptide Symposium, Gdansk, Poland, p. 238, K.Rolka, P.Rekowski, and J.Silbering, eds., Kenes, Geneva, (2007)
Catalog No. 50-211-0065