Learn More
Medchemexpress LLC Acetyl phosphate (lithium potassium) | 94249-01-1 | 98.0% | 1 ML
SDP
Catalog No. 5000340747
Encompass_Preferred
Acetyl phosphate lithium potassium is an endogenous metabolic product critical for bacterial metabolic regulation, especially in lysine acetylation. It plays a significant role in bacterial responses to environmental stress and adaptive reactions. This compound regulates lysine acetylation in *Escherichia coli*, influencing protein function, metabolic activities, and cell growth. It also non-enzymatically regulates lysine acetylation of *Salmonella typhimurium* PhoP protein, vital for bacterial responses to various environmental stresses.
  • Endogenous metabolic product vital for bacterial metabolic regulation
  • Important for bacterial responses to environmental stress and adaptation
  • Regulates lysine acetylation in *Escherichia coli*
  • Influences protein function, metabolic activities, and cell growth
  • Non-enzymatically regulates *Salmonella typhimurium* PhoP protein acetylation
Catalog No. 50-003-40747 Supplier Medchemexpress LLC Supplier No. HY12873010MM/1ML
Acetyl phosphate lithium potassium is an endogenous metabolic product critical for bacterial metabolic regulation, especially in lysine acetylation. It plays a significant role in bacterial responses to environmental stress and adaptive reactions. This compound regulates lysine acetylation in *Escherichia coli*, influencing protein function, metabolic activities, and cell growth. It also non-enzymatically regulates lysine acetylation of *Salmonella typhimurium* PhoP protein, vital for bacterial responses to various environmental stresses.
  • Endogenous metabolic product vital for bacterial metabolic regulation
  • Important for bacterial responses to environmental stress and adaptation
  • Regulates lysine acetylation in *Escherichia coli*
  • Influences protein function, metabolic activities, and cell growth
  • Non-enzymatically regulates *Salmonella typhimurium* PhoP protein acetylation