Learn More
Medchemexpress LLC Acetyl phosphate lithium potassium | 94249-01-1 | 98.0% | 10 MG
SDP
Catalog No. 5000340002
Encompass_Preferred
Acetyl phosphate lithium potassium is an endogenous metabolic product that plays a key role in bacterial metabolic regulation, particularly in lysine acetylation. It is important in bacterial responses to environmental stress and adaptive reactions.
  • Plays a key role in bacterial metabolic regulation, particularly in lysine acetylation
  • Important in bacterial responses to environmental stress and adaptive reactions
  • Regulates lysine acetylation in *Escherichia coli*, promoting protein function, influencing metabolic activities, and affecting cell growth
  • Non-enzymatically regulates lysine acetylation of *Salmonella typhimurium* PhoP protein
  • Modulates bacterial virulence via acetylation
Catalog No. 50-003-40002 Supplier Medchemexpress LLC Supplier No. HY12873010MG
Acetyl phosphate lithium potassium is an endogenous metabolic product that plays a key role in bacterial metabolic regulation, particularly in lysine acetylation. It is important in bacterial responses to environmental stress and adaptive reactions.
  • Plays a key role in bacterial metabolic regulation, particularly in lysine acetylation
  • Important in bacterial responses to environmental stress and adaptive reactions
  • Regulates lysine acetylation in *Escherichia coli*, promoting protein function, influencing metabolic activities, and affecting cell growth
  • Non-enzymatically regulates lysine acetylation of *Salmonella typhimurium* PhoP protein
  • Modulates bacterial virulence via acetylation