Learn More
Medchemexpress LLC Acetyl phosphate lithium potassium | 94249-01-1 | 98.0% | 25 MG
SDP
Catalog No. 5000342958
Encompass_Preferred
Acetyl phosphate lithium potassium is an endogenous metabolic product that plays a key role in bacterial metabolic regulation, especially in lysine acetylation. It is important for bacterial responses to environmental stress and adaptive reactions. It acts as a crucial regulator of lysine acetylation in Escherichia coli, influencing protein function, metabolic activities, and cell growth. Furthermore, it non-enzymatically regulates lysine acetylation of Salmonella typhimurium PhoP protein, which is vital for bacterial responses to various environmental stresses.
  • Endogenous metabolic product
  • Plays a key role in bacterial metabolic regulation
  • Important for bacterial responses to environmental stress and adaptive reactions
  • Regulates lysine acetylation in Escherichia coli
  • Regulates lysine acetylation of Salmonella typhimurium PhoP protein
Catalog No. 50-003-42958 Supplier Medchemexpress LLC Supplier No. 25MGHY128730
Acetyl phosphate lithium potassium is an endogenous metabolic product that plays a key role in bacterial metabolic regulation, especially in lysine acetylation. It is important for bacterial responses to environmental stress and adaptive reactions. It acts as a crucial regulator of lysine acetylation in Escherichia coli, influencing protein function, metabolic activities, and cell growth. Furthermore, it non-enzymatically regulates lysine acetylation of Salmonella typhimurium PhoP protein, which is vital for bacterial responses to various environmental stresses.
  • Endogenous metabolic product
  • Plays a key role in bacterial metabolic regulation
  • Important for bacterial responses to environmental stress and adaptive reactions
  • Regulates lysine acetylation in Escherichia coli
  • Regulates lysine acetylation of Salmonella typhimurium PhoP protein