Learn More
Medchemexpress LLC Acetyl phosphate lithium potassium | 94249-01-1 | MFCD00002134 | 98.0% | 184.06 | C2H3KLiO5P | 5 MG
SDP
Catalog No. 5000262455
Encompass_Preferred
Acetyl phosphate lithium potassium is the lithium-potassium salt of acetyl phosphate supplied as a research-grade endogenous metabolite. Provided as a dry solid with high chemical purity, it is used as a phosphate donor and acetylation reagent in biochemical and enzymatic studies. Small-scale quantities are available for low-volume laboratory experiments and assay development.
  • High purity (98.0%) suitable for research applications.
  • Available in small-scale quantities for assay development and pilot studies.
  • Soluble in water; offered as a 10 mM solution option for convenience.
  • Provides a source of acetyl phosphate for enzymatic reactions and acetylation studies.
  • Compact packaging minimizes reagent waste for low-volume experiments.
Catalog No. 50-002-62455 Supplier Medchemexpress LLC Supplier No. HY1287305MG
Acetyl phosphate lithium potassium is the lithium-potassium salt of acetyl phosphate supplied as a research-grade endogenous metabolite. Provided as a dry solid with high chemical purity, it is used as a phosphate donor and acetylation reagent in biochemical and enzymatic studies. Small-scale quantities are available for low-volume laboratory experiments and assay development.
  • High purity (98.0%) suitable for research applications.
  • Available in small-scale quantities for assay development and pilot studies.
  • Soluble in water; offered as a 10 mM solution option for convenience.
  • Provides a source of acetyl phosphate for enzymatic reactions and acetylation studies.
  • Compact packaging minimizes reagent waste for low-volume experiments.