Learn More
Medchemexpress LLC Acetyl tributyl citrate | 77-90-7 | 98.4% | 402.48 | 5 MG
SDP
Catalog No. 5000453859
Encompass_Preferred
Acetyl tributyl citrate is a pharmaceutical excipient and a biodegradable hydrophobic plasticizer. It can be utilized in cosmetics, food packaging, and as a flavoring agent for food. For research use only; not sold to patients.
  • Pharmaceutical excipient.
  • Biodegradable hydrophobic plasticizer.
  • Can be used in cosmetics.
  • Can be used in food packaging.
  • Can be used as a flavoring substance for food.
Catalog No. 50-004-53859 Supplier Medchemexpress LLC Supplier No. HYW010981S5MG
Acetyl tributyl citrate is a pharmaceutical excipient and a biodegradable hydrophobic plasticizer. It can be utilized in cosmetics, food packaging, and as a flavoring agent for food. For research use only; not sold to patients.
  • Pharmaceutical excipient.
  • Biodegradable hydrophobic plasticizer.
  • Can be used in cosmetics.
  • Can be used in food packaging.
  • Can be used as a flavoring substance for food.