Learn More
Medchemexpress LLC Acetyl tributyl citrate | 77-90-7 | 98.8% | 402.48 | 25 MG
SDP
Catalog No. 5000453249
Encompass_Preferred
Acetyl tributyl citrate (Standard) is the analytical standard of Acetyl tributyl citrate. This product is intended for research and analytical applications. It acts as an alternative plasticizer and is supplied as a reference standard assay.
  • Analytical standard for precise measurements
  • Alternative plasticizer for various applications
  • Suitable for qualitative research experiments
  • Suitable for quantitative research experiments
  • Suitable for methodological research experiments
  • Used in HPLC, GC, and MS applications
Catalog No. 50-004-53249 Supplier Medchemexpress LLC Supplier No. 25MGHYW010981R
Acetyl tributyl citrate (Standard) is the analytical standard of Acetyl tributyl citrate. This product is intended for research and analytical applications. It acts as an alternative plasticizer and is supplied as a reference standard assay.
  • Analytical standard for precise measurements
  • Alternative plasticizer for various applications
  • Suitable for qualitative research experiments
  • Suitable for quantitative research experiments
  • Suitable for methodological research experiments
  • Used in HPLC, GC, and MS applications