Learn More
Medchemexpress LLC Acetyl tributyl citrate | 77-90-7 | 98.97% | 402.48 | 10 MG
SDP
Catalog No. 5000452448
Encompass_Preferred
Acetyl tributyl citrate is a pharmaceutical excipient and biodegradable hydrophobic plasticizer. It can be used in cosmetics, food packaging, and as a flavoring substance for food. It exhibits rapid metabolism in rat plasma, rat liver microsomes, and human liver microsomes. It is also metabolized by multiple human CYP isoforms, with prominent metabolic activities from CYP2C19 and CYP3A4 at 10 μM concentration.
  • Used in pharmaceutical formulations.
  • Acts as a biodegradable plasticizer.
  • Applicable in cosmetics, food packaging, and as a food flavoring substance.
  • Exhibits rapid metabolism in rat plasma, rat liver microsomes, and human liver microsomes.
  • Metabolized by multiple human CYP isoforms, with prominent metabolic activities from CYP2C19 and CYP3A4 at 10 μM concentration.
  • In rats, it shows rapid absorption and elimination with an oral bioavailability of 27.4%.
Catalog No. 50-004-52448 Supplier Medchemexpress LLC Supplier No. HYW01098110MG
Acetyl tributyl citrate is a pharmaceutical excipient and biodegradable hydrophobic plasticizer. It can be used in cosmetics, food packaging, and as a flavoring substance for food. It exhibits rapid metabolism in rat plasma, rat liver microsomes, and human liver microsomes. It is also metabolized by multiple human CYP isoforms, with prominent metabolic activities from CYP2C19 and CYP3A4 at 10 μM concentration.
  • Used in pharmaceutical formulations.
  • Acts as a biodegradable plasticizer.
  • Applicable in cosmetics, food packaging, and as a food flavoring substance.
  • Exhibits rapid metabolism in rat plasma, rat liver microsomes, and human liver microsomes.
  • Metabolized by multiple human CYP isoforms, with prominent metabolic activities from CYP2C19 and CYP3A4 at 10 μM concentration.
  • In rats, it shows rapid absorption and elimination with an oral bioavailability of 27.4%.