Learn More
Medchemexpress LLC Acetyl tributyl citrate | 77-90-7 | 98.97% | 402.48 | 100 MG
Supplier: Medchemexpress LLC HYW010981100MG
Acetyl tributyl citrate (Tributyl O-acetylcitrate) is a pharmaceutical excipient and biodegradable hydrophobic plasticizer. It can be used in cosmetics, food packaging, and as a flavoring substance for food.
- Pharmaceutical excipient
- Biodegradable hydrophobic plasticizer
- Used in cosmetics
- Used in food packaging
- Used as a flavoring substance for food
- Exhibits rapid absorption and elimination in rats.
- Rapidly metabolized in rat plasma, rat liver microsomes, and human liver microsomes.