Learn More
Medchemexpress LLC Acetyl tributyl citrate | 77-90-7 | 99.0% | 402.48 | 25 MG
SDP
Catalog No. 5000366541
Encompass_Preferred
Acetyl tributyl citrate is a pharmaceutical excipient and biodegradable hydrophobic plasticizer. It serves various applications including use in cosmetics, food packaging, and as a food flavoring substance.
  • Used as a pharmaceutical excipient
  • Functions as a biodegradable hydrophobic plasticizer
  • Suitable for cosmetics applications
  • Appropriate for food packaging materials
  • Can be used as a food flavoring substance
  • Presents as a colorless to light yellow liquid
Catalog No. 50-003-66541 Supplier Medchemexpress LLC Supplier No. 25MGHYW010981
Acetyl tributyl citrate is a pharmaceutical excipient and biodegradable hydrophobic plasticizer. It serves various applications including use in cosmetics, food packaging, and as a food flavoring substance.
  • Used as a pharmaceutical excipient
  • Functions as a biodegradable hydrophobic plasticizer
  • Suitable for cosmetics applications
  • Appropriate for food packaging materials
  • Can be used as a food flavoring substance
  • Presents as a colorless to light yellow liquid