Learn More
Strem, An Ascensus Company CAS# 12245-39-5. 100mg. Acetylacetonato(1,5-cyclooctadiene)rhodium(I), 98%. MFCD00075046
SDP
Catalog No. 5090112041
Encompass
CAS# 12245-39-5. 100mg. Acetylacetonato(1,5-cyclooctadiene)rhodium(I), 98%. MFCD00075046. Molecular Weight: 310.19. Molecular Formula: Rh(C8H12)(C5H7O2). Color/form: orange xtl. Strem# 45-0010. http://www.strem.com/catalog/v/45-0010/
Catalog No. 50-901-12041 Supplier Strem, An Ascensus Company Supplier No. 450010100MG
CAS# 12245-39-5. 100mg. Acetylacetonato(1,5-cyclooctadiene)rhodium(I), 98%. MFCD00075046. Molecular Weight: 310.19. Molecular Formula: Rh(C8H12)(C5H7O2). Color/form: orange xtl. Strem# 45-0010. http://www.strem.com/catalog/v/45-0010/