Learn More
Strem, An Ascensus Company CAS# 12082-47-2. 500mg. Acetylacetonatobis(ethylene)rhodium(I), 99%. MFCD00015354
SDP
Catalog No. 5090112047
Encompass
CAS# 12082-47-2. 500mg. Acetylacetonatobis(ethylene)rhodium(I), 99%. MFCD00015354. Molecular Weight: 258.13. Molecular Formula: Rh(C2H4)2(C5H7O2). Color/form: yellow to orange xtl. Strem# 45-0020. http://www.strem.com/catalog/v/45-0020/
Catalog No. 50-901-12047 Supplier Strem, An Ascensus Company Supplier No. 450020500MG
CAS# 12082-47-2. 500mg. Acetylacetonatobis(ethylene)rhodium(I), 99%. MFCD00015354. Molecular Weight: 258.13. Molecular Formula: Rh(C2H4)2(C5H7O2). Color/form: yellow to orange xtl. Strem# 45-0020. http://www.strem.com/catalog/v/45-0020/