Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetylarenobufagin | 184673-79-8 | 1 MG
SDP

Catalog No. 5000417213
Encompass_Preferred

Acetylarenobufagin is a steroidal hypoxia inducible factor-1 (HIF-I) modulator. It is intended for research use only.

  • Purity of 98.63%
  • Molecular weight of 458.54
  • Chemical formula is C26H34O7
  • Appears as a solid
  • Color is white to off-white
  • Classified as a steroid
  • Initially sourced from animals (toad)
  • Targets HIF-I

Catalog No. 50-004-17213 Supplier Medchemexpress LLC Supplier No. HYN69051MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acetylarenobufagin is a steroidal hypoxia inducible factor-1 (HIF-I) modulator. It is intended for research use only.

  • Purity of 98.63%
  • Molecular weight of 458.54
  • Chemical formula is C26H34O7
  • Appears as a solid
  • Color is white to off-white
  • Classified as a steroid
  • Initially sourced from animals (toad)
  • Targets HIF-I

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.