Learn More
Cayman Chemical Acetylated Carbohydrate Mixtu
SDP
Catalog No. 502990198
Encompass_Preferred
A mixture of four pentaacetate alditols; intended for use as a reference standard for the detection of plant cell wall polysaccharides, among others, in various sample types by GC-MS; supplied in an amber ampule in which the headspace has been purged with nitrogen/argon to prevent lipid oxidation
Catalog No. 50-299-0198 Supplier Cayman Chemical Supplier No. 293671 ML
A mixture of four pentaacetate alditols; intended for use as a reference standard for the detection of plant cell wall polysaccharides, among others, in various sample types by GC-MS; supplied in an amber ampule in which the headspace has been purged with nitrogen/argon to prevent lipid oxidation