Learn More
Cayman Chemical Acetylated Carbohydrate Mixtu
SDP
Supplier:  Cayman Chemical 293681 ML
Encompass_Preferred
A mixture of four acetylated polysaccharides; intended for use as a reference standard for the detection of these polysaccharides in various sample types by GC-MS; supplied in an amber ampule in which the headspace has been purged with nitrogen/argon to prevent lipid oxidation
Catalog No. 50-298-8128