Learn More
Sino Biological Peptide Substrates: Acetylated Histone H3 Peptide I
Catalog No. 502569046
Encompass_Preferred
The Acetylated Histone H3 (K9, 14) Peptide sequence (ARTKQTAR[Ac-K]STGG[Ac-K]APRKQLAGGKKC) is derived from human histone H3 (1-21) and is suitable for use as the assay control of histone (de-)methylation and (de-) acetylation.
Catalog No. 50-256-9046 Supplier Sino Biological Supplier No. H12358500
The Acetylated Histone H3 (K9, 14) Peptide sequence (ARTKQTAR[Ac-K]STGG[Ac-K]APRKQLAGGKKC) is derived from human histone H3 (1-21) and is suitable for use as the assay control of histone (de-)methylation and (de-) acetylation.