Learn More
Cayman Chemical β-acetyl-Boswellic Acid, Purity: ≥95%, CAS: 5968-70-7, 10 mg
SDP
Catalog No. 502952242
Encompass_Preferred
A non-reducing inhibitor of 5-LO that shows selectivity for 5-LO over 12-LO or COX; inhibits topoisomerase II activity and arrests cell cycling or induces apoptosis in cancer cells
Catalog No. 50-295-2242 Supplier Cayman Chemical Supplier No. 1167410 MG
A non-reducing inhibitor of 5-LO that shows selectivity for 5-LO over 12-LO or COX; inhibits topoisomerase II activity and arrests cell cycling or induces apoptosis in cancer cells