Learn More
Eurofins Discoverx Acetylcholine, 10 mg. (92-1125)
Supplier:  Eurofins Discoverx 921125
Encompass
Acetylcholine is a neurotransmitter of the autonomic nervous system and is the principle neurotransmitter of the somatic nervous system and autonomic ganglia. It is the most common neurotransmitter in the body, and acts as a neuromodulator of the CNS and PNS, having effects on attention, arousal, and muscle activation. Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9658
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.