Learn More
Medchemexpress LLC Acetylcholine bromide | 66-23-9 | 99.02% | 10 G
SDP
Catalog No. 5000406606
Encompass_Preferred
Acetylcholine bromide is an organic choline salt compound utilized in neurophysiology and pharmacology experiments. It acts as a neurotransmitter in the nervous system, transmitting signals and regulating various essential physiological functions. Its distinct biological activity makes it valuable for research concerning neurotransmitters, nerve damage, and muscle movement.
  • Organic choline salt compound
  • Used in neurophysiology and pharmacology experiments
  • Functions as a neurotransmitter
  • Regulates important physiological functions
  • Widely used in studies of neurotransmitters, nerve damage, and muscle movement
  • Biochemical reagent for life science related research
Catalog No. 50-004-06606 Supplier Medchemexpress LLC Supplier No. HYB0282A10G
Acetylcholine bromide is an organic choline salt compound utilized in neurophysiology and pharmacology experiments. It acts as a neurotransmitter in the nervous system, transmitting signals and regulating various essential physiological functions. Its distinct biological activity makes it valuable for research concerning neurotransmitters, nerve damage, and muscle movement.
  • Organic choline salt compound
  • Used in neurophysiology and pharmacology experiments
  • Functions as a neurotransmitter
  • Regulates important physiological functions
  • Widely used in studies of neurotransmitters, nerve damage, and muscle movement
  • Biochemical reagent for life science related research