Learn More
Medchemexpress LLC Acetylcholine bromide (ACh bromide) | 66-23-9 | 99.0% | 5 G
SDP
Catalog No. 5000408716
Encompass_Preferred
Acetylcholine bromide is an organic choline salt compound that finds common application in neurophysiology and pharmacology experiments. It is crucial in the nervous system, where it acts as a neurotransmitter, facilitating signal transmission and regulating various vital physiological functions. Its distinct biological activity makes it widely used in research concerning neurotransmitters, nerve damage, and muscle movement.
  • Used as a biochemical reagent in life science research.
  • Plays a vital role in nervous system signal transmission.
  • Regulates important physiological functions.
  • Utilized in studies related to neurotransmitters.
  • Applicable in research on nerve damage and muscle movement.
Catalog No. 50-004-08716 Supplier Medchemexpress LLC Supplier No. HYB0282A5G
Acetylcholine bromide is an organic choline salt compound that finds common application in neurophysiology and pharmacology experiments. It is crucial in the nervous system, where it acts as a neurotransmitter, facilitating signal transmission and regulating various vital physiological functions. Its distinct biological activity makes it widely used in research concerning neurotransmitters, nerve damage, and muscle movement.
  • Used as a biochemical reagent in life science research.
  • Plays a vital role in nervous system signal transmission.
  • Regulates important physiological functions.
  • Utilized in studies related to neurotransmitters.
  • Applicable in research on nerve damage and muscle movement.