Learn More
Sigma Aldrich Fine Chemicals Biosciences Acetylcholine chloride, 60-31-1, MFCD00011698, 25 G
Catalog No. NC0550047
Encompass
Linear Formula: (CH3)3N+CH2CH2OCOCH3Cl-, Molecular Weight: 181.66, ≥99% (TLC), Synonym: Ach.
Catalog No. NC0550047 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A662525G
Linear Formula: (CH3)3N+CH2CH2OCOCH3Cl-, Molecular Weight: 181.66, ≥99% (TLC), Synonym: Ach.