Learn More
Apexbio Technology LLC Acetylcholine Chloride 60-31-1 1g
SDP
Catalog No. 5000091262
Encompass_Preferred
Acetylcholine chloride is an endogenous cholinergic agonist targeting nicotinic and muscarinic acetylcholine receptors It is designed to modulate cholinergic neurotransmission thereby regulating muscular contraction autonomic signaling and neuroendocrine responses Acetylcholine chloride exerts its biological activity primarily through binding and activating specific cholinergic receptors In experimental studies acetylcholine chloride demonstrates receptor activation properties and is frequently utilized in vitro to investigate receptor-ligand interactions synaptic transmission and neural transduction pathways Based on these pharmacological properties acetylcholine chloride holds research potential in neuroscience and pharmacological studies focused on cholinergic signaling and receptor characterization
Catalog No. 50-000-91262 Supplier Apexbio Technology LLC Supplier No. B15961000
Acetylcholine chloride is an endogenous cholinergic agonist targeting nicotinic and muscarinic acetylcholine receptors It is designed to modulate cholinergic neurotransmission thereby regulating muscular contraction autonomic signaling and neuroendocrine responses Acetylcholine chloride exerts its biological activity primarily through binding and activating specific cholinergic receptors In experimental studies acetylcholine chloride demonstrates receptor activation properties and is frequently utilized in vitro to investigate receptor-ligand interactions synaptic transmission and neural transduction pathways Based on these pharmacological properties acetylcholine chloride holds research potential in neuroscience and pharmacological studies focused on cholinergic signaling and receptor characterization