Learn More
Medchemexpress LLC Acetylcholine-d13 bromide | 66-23-9 | 98.6% | 5 MG
SDP
Catalog No. 5000406064
Encompass_Preferred
Acetylcholine-d13 bromide is a deuterium-labeled form of Acetylcholine bromide. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs. This product is intended for research use only.
  • Can be used as a tracer
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Contains stable heavy isotopes incorporated into drug molecules for quantitation during drug development
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-06064 Supplier Medchemexpress LLC Supplier No. HYB0282AS25MG
Acetylcholine-d13 bromide is a deuterium-labeled form of Acetylcholine bromide. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs. This product is intended for research use only.
  • Can be used as a tracer
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Contains stable heavy isotopes incorporated into drug molecules for quantitation during drug development
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs