Learn More
Medchemexpress LLC Acetylcholine-d16 bromide | 347841-43-4 | 98.6% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0282AS31MG
Encompass_Preferred
Acetylcholine-d16 (bromide) is a deuterium labeled Acetylcholine bromide intended for research use. It serves as a valuable tool for quantitative analysis using techniques like NMR, GC-MS, or LC-MS, where it can act as a tracer or an internal standard.
  • Stable heavy isotopes are incorporated into drug molecules
  • Used as tracers for quantitation during drug development
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-06475