Learn More
Medchemexpress LLC Acetylcholine-d4 (bromide) | 93449-31-1 | 98.0% | 10 MG
SDP
Catalog No. 5000298272
Encompass_Preferred
Acetylcholine-d4 (bromide) is the deuterium-labeled analogue of acetylcholine bromide supplied for research and analytical use. It serves as an isotopically labeled tracer and internal standard for quantitative assays and is provided as a high-purity solid suitable for spectroscopic and mass-spectrometric workflows.
  • isotopically labeled internal standard for quantitative analysis
  • compatible with NMR, GC-MS, and LC-MS workflows
  • high chemical purity (98.0%) for reliable quantitation
  • solid, white to off-white appearance for ease of handling
  • stable when stored sealed and protected from moisture; in solvent: -80°C (up to 6 months) or -20°C (up to 1 month)
  • intended for research use only
Catalog No. 50-002-98272 Supplier Medchemexpress LLC Supplier No. HYB0282AS110MG
Acetylcholine-d4 (bromide) is the deuterium-labeled analogue of acetylcholine bromide supplied for research and analytical use. It serves as an isotopically labeled tracer and internal standard for quantitative assays and is provided as a high-purity solid suitable for spectroscopic and mass-spectrometric workflows.
  • isotopically labeled internal standard for quantitative analysis
  • compatible with NMR, GC-MS, and LC-MS workflows
  • high chemical purity (98.0%) for reliable quantitation
  • solid, white to off-white appearance for ease of handling
  • stable when stored sealed and protected from moisture; in solvent: -80°C (up to 6 months) or -20°C (up to 1 month)
  • intended for research use only