Learn More
Medchemexpress LLC Acetylcholine-d4 bromide | 93449-31-1 | 98.69% | 5 MG
SDP
Catalog No. 5000405966
Encompass_Preferred
Acetylcholine-d4 (bromide) is the deuterium labeled Acetylcholine bromide. This compound is used as a tracer and an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development, and deuteration may affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Intended for research use only
  • Not for sale to patients
Catalog No. 50-004-05966 Supplier Medchemexpress LLC Supplier No. HYB0282AS15MG
Acetylcholine-d4 (bromide) is the deuterium labeled Acetylcholine bromide. This compound is used as a tracer and an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development, and deuteration may affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Intended for research use only
  • Not for sale to patients