Learn More
Medchemexpress LLC Acetylcholine-d4 chloride | 344298-94-8 | 99.21% | 5 MG
SDP
Catalog No. 5000405832
Encompass_Preferred
Acetylcholine-d4 (chloride) is the deuterium labeled Acetylcholine chloride. Acetylcholine chloride (ACh chloride) is a neurotransmitter, a potent and BBB-permeable cholinergic agonist, and a modulator of the activity of dopaminergic (DAergic) neurons through the stimulation of nicotinic acetylcholine receptors (nAChRs). It also inhibits p53 mutant peptide aggregation in vitro.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes have potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-05832 Supplier Medchemexpress LLC Supplier No. HYB0282S5MG
Acetylcholine-d4 (chloride) is the deuterium labeled Acetylcholine chloride. Acetylcholine chloride (ACh chloride) is a neurotransmitter, a potent and BBB-permeable cholinergic agonist, and a modulator of the activity of dopaminergic (DAergic) neurons through the stimulation of nicotinic acetylcholine receptors (nAChRs). It also inhibits p53 mutant peptide aggregation in vitro.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes have potential to affect pharmacokinetic and metabolic profiles of drugs