Learn More
Medchemexpress LLC Acetylcholine-d9 bromide | 93449-32-2 | 98.9% | 1 MG
SDP
Catalog No. 5000408717
Encompass_Preferred
Acetylcholine-d9 (bromide) is the deuterium labeled Acetylcholine bromide.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-08717 Supplier Medchemexpress LLC Supplier No. HYB0282AS1MG
Acetylcholine-d9 (bromide) is the deuterium labeled Acetylcholine bromide.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.