Learn More
Medchemexpress LLC Acetylcholine-d9 bromide | 93449-32-2 | 98.9% | 5 MG
SDP
Catalog No. 5000406063
Encompass_Preferred
Acetylcholine-d9 bromide is a deuterium labeled Acetylcholine bromide, commonly used for quantitative analysis in drug development.
  • Used as a tracer.
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes like deuterium.
  • Potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-06063 Supplier Medchemexpress LLC Supplier No. HYB0282AS5MG
Acetylcholine-d9 bromide is a deuterium labeled Acetylcholine bromide, commonly used for quantitative analysis in drug development.
  • Used as a tracer.
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes like deuterium.
  • Potential to affect pharmacokinetic and metabolic profiles of drugs.