Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetylcholine-d9 bromide | 93449-32-2 | MFCD01073946 | 98.9% | 235.17 | C7H7D9BrNO2 | 10 MG
SDP

Catalog No. 5000297076
Encompass_Preferred

Acetylcholine-d9 bromide is the deuterium-labeled analogue of acetylcholine bromide used as a stable isotope standard or internal reference for quantitative bioanalytical methods such as LC-MS. It is supplied as a hygroscopic ionic salt in small milligram quantities for use in method development, calibration, and tracer studies.

  • Deuterium-labeled internal standard for quantitative analysis.
  • High purity suitable for analytical workflows.
  • Supplied in small milligram quantities for trace-level assays.
  • Molecular formula C7H7D9BrNO2; molecular weight 235.17.
  • Store sealed at 4°C and protect from moisture.

Catalog No. 50-002-97076 Supplier Medchemexpress LLC Supplier No. HYB0282AS10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acetylcholine-d9 bromide is the deuterium-labeled analogue of acetylcholine bromide used as a stable isotope standard or internal reference for quantitative bioanalytical methods such as LC-MS. It is supplied as a hygroscopic ionic salt in small milligram quantities for use in method development, calibration, and tracer studies.

  • Deuterium-labeled internal standard for quantitative analysis.
  • High purity suitable for analytical workflows.
  • Supplied in small milligram quantities for trace-level assays.
  • Molecular formula C7H7D9BrNO2; molecular weight 235.17.
  • Store sealed at 4°C and protect from moisture.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.