Learn More
Medchemexpress LLC Acetylcholine-d9 chloride | 344298-95-9 | 99.0% | 1 MG
SDP
Catalog No. 5000407021
Encompass_Preferred
Acetylcholine-d9 chloride is a deuterium-labeled acetylcholine chloride, which is a potent, BBB-permeable cholinergic agonist. This neurotransmitter modulates the activity of dopaminergic neurons by stimulating nicotinic acetylcholine receptors and inhibits p53 mutant peptide aggregation in vitro. It can be utilized as a tracer or an internal standard for quantitative analysis through techniques like NMR, GC-MS, or LC-MS.
  • Deuterium-labeled acetylcholine chloride
  • Potent and BBB-permeable cholinergic agonist
  • Modulates dopaminergic neuron activity
  • Inhibits p53 mutant peptide aggregation in vitro
  • Suitable as a tracer or internal standard for quantitative analysis
Catalog No. 50-004-07021 Supplier Medchemexpress LLC Supplier No. HYB0282S11MG
Acetylcholine-d9 chloride is a deuterium-labeled acetylcholine chloride, which is a potent, BBB-permeable cholinergic agonist. This neurotransmitter modulates the activity of dopaminergic neurons by stimulating nicotinic acetylcholine receptors and inhibits p53 mutant peptide aggregation in vitro. It can be utilized as a tracer or an internal standard for quantitative analysis through techniques like NMR, GC-MS, or LC-MS.
  • Deuterium-labeled acetylcholine chloride
  • Potent and BBB-permeable cholinergic agonist
  • Modulates dopaminergic neuron activity
  • Inhibits p53 mutant peptide aggregation in vitro
  • Suitable as a tracer or internal standard for quantitative analysis