Learn More
Apexbio Technology LLC ACETYLCHOLINE-IODIDE-200MG
SDP
Catalog No. 5000569293
Encompass_Preferred
Acetylcholine iodide (CAS No 2260-50-6) is a chemical compound used extensively in biomedical research to study the cholinergic system which plays a crucial role in neurotransmission Originating from the acetylcholine family it functions by mimicking acetylcholine s action on muscarinic and nicotinic receptors influencing various physiological responses In vitro its activity is often observed in a nanomolar to low micromolar range making it suitable for probing cellular responses Researchers commonly use acetylcholine iodide in cell models and animal studies to investigate synaptic functions receptor dynamics and potential therapeutic effects It is also employed in drug screening assays to explore interactions with cholinergic targets Experimental concentrations typically depend on the specific research design ensuring appropriate contextual application
Catalog No. 50-005-69293 Supplier Apexbio Technology LLC Supplier No. C6791200
Acetylcholine iodide (CAS No 2260-50-6) is a chemical compound used extensively in biomedical research to study the cholinergic system which plays a crucial role in neurotransmission Originating from the acetylcholine family it functions by mimicking acetylcholine s action on muscarinic and nicotinic receptors influencing various physiological responses In vitro its activity is often observed in a nanomolar to low micromolar range making it suitable for probing cellular responses Researchers commonly use acetylcholine iodide in cell models and animal studies to investigate synaptic functions receptor dynamics and potential therapeutic effects It is also employed in drug screening assays to explore interactions with cholinergic targets Experimental concentrations typically depend on the specific research design ensuring appropriate contextual application