Learn More
Medchemexpress LLC Acetylcholine iodide | 2260-50-6 | MFCD00011815 | 98.6% | 273.11 g/mol | C7H16INO2 | 1 ML
SDP
Catalog No. 5000213344
Encompass_Preferred
Acetylcholine iodide is the iodide salt of the neurotransmitter acetylcholine, supplied as a research reagent for receptor pharmacology and physiological studies. It is offered as a 10 mM solution in DMSO (1 mL) or as a solid; common applications include muscarinic and nicotinic receptor assays, studies of intestinal ion transport, and mammary gland physiology research.
  • Ready-to-use 10 mM solution in DMSO (1 mL) for rapid assay setup.
  • Also available as a solid for custom concentration and formulation.
  • High purity suitable for biochemical and pharmacological assays.
  • Comprehensive documentation: datasheet, SDS, COA, and spectral data.
  • Molecular weight 273.11 g/mol; CAS number 2260-50-6 for clear identification.
Catalog No. 50-002-13344 Supplier Medchemexpress LLC Supplier No. HY10108610MM 1ML
Acetylcholine iodide is the iodide salt of the neurotransmitter acetylcholine, supplied as a research reagent for receptor pharmacology and physiological studies. It is offered as a 10 mM solution in DMSO (1 mL) or as a solid; common applications include muscarinic and nicotinic receptor assays, studies of intestinal ion transport, and mammary gland physiology research.
  • Ready-to-use 10 mM solution in DMSO (1 mL) for rapid assay setup.
  • Also available as a solid for custom concentration and formulation.
  • High purity suitable for biochemical and pharmacological assays.
  • Comprehensive documentation: datasheet, SDS, COA, and spectral data.
  • Molecular weight 273.11 g/mol; CAS number 2260-50-6 for clear identification.