Learn More
Selleck Chemical LLC Acetylcholine iodide S4718-100mg
SDP
Supplier:  Selleck Chemical LLC S4718100MG
Encompass_Preferred
Acetylcholine iodide (Acetylcolina) is a neurotransmitter found at neuromuscular junctions autonomic ganglia parasympathetic effector junctions a subset of sympathetic effector junctions and at many sites in the central nervous system
Catalog No. 50-310-9908