Learn More
Selleck Chemical LLC Acetylcholine iodide S4718-500mg
SDP
Catalog No. 503112052
Encompass_Preferred
Acetylcholine iodide (Acetylcolina) is a neurotransmitter found at neuromuscular junctions autonomic ganglia parasympathetic effector junctions a subset of sympathetic effector junctions and at many sites in the central nervous system
Catalog No. 50-311-2052 Supplier Selleck Chemical LLC Supplier No. S4718500MG
Acetylcholine iodide (Acetylcolina) is a neurotransmitter found at neuromuscular junctions autonomic ganglia parasympathetic effector junctions a subset of sympathetic effector junctions and at many sites in the central nervous system