Learn More
Cusabio Technology LLC Acetylcholinesterase AChE
Catalog No. 5000050058
Encompass_Preferred
This kit detects AChE (Acetylcholinesterase) in mouse serum plasma cell culture supernates and tissue homogenates using a quantitative sandwich ELISA method It offers a detection range of 0 312-20 mU/mL with a sensitivity of 0 078 mU/mL providing precise measurement of AChE concentrations in biological samples The assay requires 50-100 L of sample volume and can be completed within 1-5 hours making it suitable for efficient laboratory workflows Detection is performed at 450 nm wavelength ensuring reliable optical measurement Designed for research applications this kit maintains consistent performance across multiple sample types while adhering to standard ELISA protocols for accuracy and reproducibility in acetylcholinesterase level quantification
Catalog No. 50-000-50058 Supplier Cusabio Technology LLC Supplier No. CSBE17521M
This kit detects AChE (Acetylcholinesterase) in mouse serum plasma cell culture supernates and tissue homogenates using a quantitative sandwich ELISA method It offers a detection range of 0 312-20 mU/mL with a sensitivity of 0 078 mU/mL providing precise measurement of AChE concentrations in biological samples The assay requires 50-100 L of sample volume and can be completed within 1-5 hours making it suitable for efficient laboratory workflows Detection is performed at 450 nm wavelength ensuring reliable optical measurement Designed for research applications this kit maintains consistent performance across multiple sample types while adhering to standard ELISA protocols for accuracy and reproducibility in acetylcholinesterase level quantification