Learn More
Sigma Aldrich Fine Chemicals Biosciences Acetylcholinesterase Activ
Catalog No. 502901917
Encompass
The Acetylcholinesterase Activity Assay Kit (Colorimetric) provides a simple quick and direct procedure for measuring Acetylcholinesterase levels in a variety of samples such as serum plasma tissues or cell lysates / cell extracts. This assay is based on the Ellman method in which thiocholine produced by Acetylcholinesterase reacts with DTNB 5 5A¢-dithiobis (2-nitrobenzoic acid) to form a colorimetric product (412 nm) proportional to the Acetylcholinesterase activity present. This kit does not require weighing or mixing of multiple reagents. The Acetylcholinesterase activity is measured without the use of a calibration curve. The Acetylcholinesterase Positive Control in the kit can be used to screen for Acetylcholinesterase inhibitors. The detection range 10-600 units/liter of Acetylcholinesterase activity.The detection time 5-10 minutesAcetylcholinesterase ( AChE) is a serine hydrolase found at neuromuscular junctions and cholinergic brain synapses. AChE hydrolyzes choline es
Catalog No. 50-290-1917 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CS00031KT
The Acetylcholinesterase Activity Assay Kit (Colorimetric) provides a simple quick and direct procedure for measuring Acetylcholinesterase levels in a variety of samples such as serum plasma tissues or cell lysates / cell extracts. This assay is based on the Ellman method in which thiocholine produced by Acetylcholinesterase reacts with DTNB 5 5A¢-dithiobis (2-nitrobenzoic acid) to form a colorimetric product (412 nm) proportional to the Acetylcholinesterase activity present. This kit does not require weighing or mixing of multiple reagents. The Acetylcholinesterase activity is measured without the use of a calibration curve. The Acetylcholinesterase Positive Control in the kit can be used to screen for Acetylcholinesterase inhibitors. The detection range 10-600 units/liter of Acetylcholinesterase activity.The detection time 5-10 minutesAcetylcholinesterase ( AChE) is a serine hydrolase found at neuromuscular junctions and cholinergic brain synapses. AChE hydrolyzes choline es