Learn More
Arbor Assays Acetylcholinesterase Fluorescent Activity Kit (2 Plate)
SDP
Supplier:  Arbor Assays K015F1
Encompass_Preferred
The DetectX Acetylcholinesterase (AChE) Activity Kit is designed to quantitatively measure AChE activity in a variety of samples, utilizing a proprietary non-fluorescent molecule, ThioStar, which fluoresces when bound to the thiol reaction product of AChE and its substrate. 
Catalog No. 50-227-8404