Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Acetylcholinesterase Fluorescent Activity Kit

Catalog No. EIAACHEF
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EIAACHEF 2 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EIAACHEF Supplier Invitrogen™ Supplier No. EIAACHEF
Add to Cart
Add to Cart

Includes

96-well plates, standard, detection reagent, assay buffer, substrate, and lot-specific technical data sheet.

Assay Acetylcholinesterase Activity in Serum, Plasma, and Erythrocyte Membrane samples.

The Acetylcholinesterase Fluorescent Activity research-use-only kit is a fluorescent activity assay designed for the quantification and detection of acetylcholinesterase activity in serum, plasma, and erythrocyte membrane samples.

This complete, ready-to-use kit includes black 96-well plate(s), acetylcholinesterase standard (1,000 mU/mL), acetylcholinesterase substrate, acetylthiocholine iodide, and other components to perform the assay. A fluorescence 96-well microplate reader capable of reading fluorescent emission at 510 nm, with excitation at 390 nm, is required for use of this kit.

Performance characteristics
• Assay type: fluorescent activity kit
• Sample types: serum, plasma, and erythrocyte membrane samples
• Sensitivity: 0.218 mU/mL
• Standard curve range: 6.25–100 mU/mL
• Reactivity: species independent

Background
Acetylcholine (ACh) is an essential neurotransmitter in the central and peripheral nervous systems. In the brain multiple areas exist where cholinergic neurons are concentrated. Nicotinic and muscarinic ACh receptors are recognized as binding and effector proteins to mediate chemical neurotransmission at neurons, ganglia, heart, and smooth muscle fibers and glands.

Acetylcholinesterase is encoded by the single AChE gene; the structural diversity in the gene products arises from alternative mRNA splicing and post translational associations of catalytic and structural subunits. The major form of acetylcholinesterase found in brain, muscle, and other tissues is the hydrophilic species, which forms disulfide-linked oligomers with collagenous, lipid-containing structural subunits. The other alternatively-spliced form, expressed primarily in the erythroid tissues, is structurally different at the C-terminal end and contains a cleavable peptide with a GPI anchor. It associates with membrane receptors through phosphoinositide moieties added post-translationally. This assay has been validated for serum, EDTA and heparin plasma, and solubilized RBC ghosts from a variety of species.

Assay principle
The Acetylcholinesterase Activity kit is designed to quantitatively measure acetylcholinesterase activity in a variety of samples. A human AChE standard is provided to generate a standard curve for the assay and all samples should be read off the standard curve. The kit utilizes a proprietary non-fluorescent molecule, that covalently binds to the thiol product of the reaction between the AChE Substrate and AChE in the standards or samples, yielding a fluorescent product read at 510 nm in a fluorescent plate reader with excitation at 390 nm. The kit is suitable for measuring AchE activity in appropriately diluted serum, plasma, and RBC ghosts from a number of species. It will also measure AChE in extracted tissue samples and cell lysates.

AChE unit definition
One unit will hydrolyze 1.0 μmol of acetylthiocholine iodide per minute at pH 7.4 and 37°C.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 96-well plate(s), standard, detection reagent, assay buffer, substrate, and lot-specific technical data sheet.

Store at –20°C.
Immunoassay Kit Format Assay Activity Kit
Sample Type Plasma, Serum
For Use With (Equipment) Microplate Reader
Quantity 2 x 96 Tests
Shipping Condition Approved for shipment on Wet or Dry Ice
Target Kit Named Acetylcholinesterase
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.