Learn More
Sigma Aldrich Fine Chemicals Biosciences Acetylcholinesterase Inhib
Catalog No. 502904296
Encompass
Acetylcholinesterase (AChE) also known as RBC cholinesterase is found primarily in the blood and neural synapses. AChE catalyzes the hydrolysis of the neurotransmitter acetylcholine into choline and acetic acid a reaction necessary to allow a cholinergic neuron to return to its resting state after activation. Inhibition of the enzyme leads to acetylcholine accumulation hyperstimulation of nicotinic and muscarinic receptors and disrupted neurotransmission. AChE inhibition is an important target for the management of Alzheimers disease and AChE inhibitors are the most common drugs used for its management. In addition to Alzheimers disease AChE inhibitors have been useful in the diagnosis or treatment of diseases such as glaucoma myasthenia gravis bladder distention and more. The Acetylcholinesterase Inhibitor Screening kit is based on an improved Ellman method in which thiocholine produced by the action of acetylcholinesterase forms a yellow color with 5 5-dithiobis(2-nitrobenz
Catalog No. 50-290-4296 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MAK3241KT
Acetylcholinesterase (AChE) also known as RBC cholinesterase is found primarily in the blood and neural synapses. AChE catalyzes the hydrolysis of the neurotransmitter acetylcholine into choline and acetic acid a reaction necessary to allow a cholinergic neuron to return to its resting state after activation. Inhibition of the enzyme leads to acetylcholine accumulation hyperstimulation of nicotinic and muscarinic receptors and disrupted neurotransmission. AChE inhibition is an important target for the management of Alzheimers disease and AChE inhibitors are the most common drugs used for its management. In addition to Alzheimers disease AChE inhibitors have been useful in the diagnosis or treatment of diseases such as glaucoma myasthenia gravis bladder distention and more. The Acetylcholinesterase Inhibitor Screening kit is based on an improved Ellman method in which thiocholine produced by the action of acetylcholinesterase forms a yellow color with 5 5-dithiobis(2-nitrobenz