Learn More
Cayman Chemical AcetylCoenzyme AsodIM sal 10mg
SDP
Catalog No. 502973248
Encompass_Preferred
A pivotal molecule in the Krebs cycle, in the synthesis of fatty acids, and in isoprenoid-based protein modifications; serves as an intermediate in oxidation of fatty acids and amino acids; an essential cofactor or substrate for acetyltransferases and acyltransferases
Catalog No. 50-297-3248 Supplier Cayman Chemical Supplier No. 1616010 MG
A pivotal molecule in the Krebs cycle, in the synthesis of fatty acids, and in isoprenoid-based protein modifications; serves as an intermediate in oxidation of fatty acids and amino acids; an essential cofactor or substrate for acetyltransferases and acyltransferases